Learn More
Hampton Research CRYSTAL SCREEN LITE NO01 185ML
SDP
Catalog No. 501606
Encompass
HR2-916-01/Crystal Screen Lite #01/MembFac HT (E01), 185 ml - 0.02 M Calcium chloride dihydrate, 0.1 M Sodium acetate trihydrate pH 4.6, 15% v/v (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.
Catalog No. 50-160-6 Supplier Hampton Research Supplier No. HR291601
HR2-916-01/Crystal Screen Lite #01/MembFac HT (E01), 185 ml - 0.02 M Calcium chloride dihydrate, 0.1 M Sodium acetate trihydrate pH 4.6, 15% v/v (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.