Learn More
Hampton Research CRYSTAL SCREEN LITE NO05 185ML
SDP
Catalog No. 501610
Encompass
HR2-916-05/Crystal Screen Lite #05/MembFac HT (E05), 185 ml - 0.2 M Sodium citrate tribasic dihydrate, 0.1 M HEPES sodium pH 7.5, 15% v/v (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.
Catalog No. 50-161-0 Supplier Hampton Research Supplier No. HR291605
HR2-916-05/Crystal Screen Lite #05/MembFac HT (E05), 185 ml - 0.2 M Sodium citrate tribasic dihydrate, 0.1 M HEPES sodium pH 7.5, 15% v/v (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.