Learn More
Hampton Research CRYSTAL SCREEN LITE NO06 185ML
SDP
Catalog No. 501611
Encompass
HR2-916-06/Crystal Screen Lite #06/MembFac HT (E06), 185 ml - 0.2 M Magnesium chloride hexahydrate, 0.1 M TRIS hydrochloride pH 8.5, 15% w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.
Catalog No. 50-161-1 Supplier Hampton Research Supplier No. HR291606
HR2-916-06/Crystal Screen Lite #06/MembFac HT (E06), 185 ml - 0.2 M Magnesium chloride hexahydrate, 0.1 M TRIS hydrochloride pH 8.5, 15% w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.