Learn More
Hampton Research CRYSTAL SCREEN LITE NO07 185ML
SDP
Catalog No. 501612
Encompass
HR2-916-07/Crystal Screen Lite #07/MembFac HT (E07), 185 ml - 0.1 M Sodium cacodylate trihydrate pH 6.5, 0.7 M Sodium acetate trihydrate. Please note items from this supplier are non-returnable.
Catalog No. 50-161-2 Supplier Hampton Research Supplier No. HR291607
HR2-916-07/Crystal Screen Lite #07/MembFac HT (E07), 185 ml - 0.1 M Sodium cacodylate trihydrate pH 6.5, 0.7 M Sodium acetate trihydrate. Please note items from this supplier are non-returnable.