Learn More
Hampton Research CRYSTAL SCREEN LITE NO08 185ML
SDP
Catalog No. 501613
Encompass
HR2-916-08/Crystal Screen Lite #08/MembFac HT (E08), 185 ml - 0.2 M Sodium citrate tribasic dihydrate, 0.1 M Sodium cacodylate trihydrate pH 6.5, 15% v/v 2-Propanol. Please note items from this supplier are non-returnable.
Catalog No. 50-161-3 Supplier Hampton Research Supplier No. HR291608
HR2-916-08/Crystal Screen Lite #08/MembFac HT (E08), 185 ml - 0.2 M Sodium citrate tribasic dihydrate, 0.1 M Sodium cacodylate trihydrate pH 6.5, 15% v/v 2-Propanol. Please note items from this supplier are non-returnable.