Learn More
Hampton Research CRYSTAL SCREEN LITE NO09 185ML
SDP
Catalog No. 501614
Encompass
HR2-916-09/Crystal Screen Lite #09/MembFac HT (E09), 185 ml - 0.2 M Ammonium acetate, 0.1 M Sodium citrate tribasic dihydrate pH 5.6, 15% w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.
Catalog No. 50-161-4 Supplier Hampton Research Supplier No. HR291609
HR2-916-09/Crystal Screen Lite #09/MembFac HT (E09), 185 ml - 0.2 M Ammonium acetate, 0.1 M Sodium citrate tribasic dihydrate pH 5.6, 15% w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.