Learn More
Hampton Research CRYSTAL SCREEN LITE NO10 185ML
SDP
Catalog No. 501615
Encompass
HR2-916-10/Crystal Screen Lite #10/MembFac HT (E10), 185 ml - 0.2 M Ammonium acetate, 0.1 M Sodium acetate trihydrate pH 4.6, 15% w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.
Catalog No. 50-161-5 Supplier Hampton Research Supplier No. HR291610
HR2-916-10/Crystal Screen Lite #10/MembFac HT (E10), 185 ml - 0.2 M Ammonium acetate, 0.1 M Sodium acetate trihydrate pH 4.6, 15% w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.