Learn More
Hampton Research CRYSTAL SCREEN LITE NO11 185ML
SDP
Catalog No. 501616
Encompass
HR2-916-11/Crystal Screen Lite #11/MembFac HT (E11), 185 ml - 0.1 M Sodium citrate tribasic dihydrate pH 5.6, 0.5 M Ammonium phosphate monobasic. Please note items from this supplier are non-returnable.
Catalog No. 50-161-6 Supplier Hampton Research Supplier No. HR291611
HR2-916-11/Crystal Screen Lite #11/MembFac HT (E11), 185 ml - 0.1 M Sodium citrate tribasic dihydrate pH 5.6, 0.5 M Ammonium phosphate monobasic. Please note items from this supplier are non-returnable.