Learn More
Hampton Research CRYSTAL SCREEN LITE NO12 185ML
SDP
Catalog No. 501617
Encompass
HR2-916-12/Crystal Screen Lite #12/MembFac HT (E12), 185 ml - 0.2 M Magnesium chloride hexahydrate, 0.1 M HEPES sodium pH 7.5, 15% v/v 2-Propanol. Please note items from this supplier are non-returnable.
Catalog No. 50-161-7 Supplier Hampton Research Supplier No. HR291612
HR2-916-12/Crystal Screen Lite #12/MembFac HT (E12), 185 ml - 0.2 M Magnesium chloride hexahydrate, 0.1 M HEPES sodium pH 7.5, 15% v/v 2-Propanol. Please note items from this supplier are non-returnable.