Learn More
Hampton Research CRYSTAL SCREEN LITE NO14 185ML
SDP
Catalog No. 501619
Encompass
HR2-916-14/Crystal Screen Lite #14/MembFac HT (F02), 185 ml - 0.2 M Calcium chloride dihydrate, 0.1 M HEPES sodium pH 7.5, 14% v/v Polyethylene glycol 400. Please note items from this supplier are non-returnable.
Catalog No. 50-161-9 Supplier Hampton Research Supplier No. HR291614
HR2-916-14/Crystal Screen Lite #14/MembFac HT (F02), 185 ml - 0.2 M Calcium chloride dihydrate, 0.1 M HEPES sodium pH 7.5, 14% v/v Polyethylene glycol 400. Please note items from this supplier are non-returnable.