Learn More
Hampton Research CRYSTAL SCREEN LITE NO15 185ML
SDP
Catalog No. 501620
Encompass
HR2-916-15/Crystal Screen Lite #15/MembFac HT (F03), 185 ml - 0.2 M Ammonium sulfate, 0.1 M Sodium cacodylate trihydrate pH 6.5, 15% w/v Polyethylene glycol 8,000. Please note items from this supplier are non-returnable.
Catalog No. 50-162-0 Supplier Hampton Research Supplier No. HR291615
HR2-916-15/Crystal Screen Lite #15/MembFac HT (F03), 185 ml - 0.2 M Ammonium sulfate, 0.1 M Sodium cacodylate trihydrate pH 6.5, 15% w/v Polyethylene glycol 8,000. Please note items from this supplier are non-returnable.