Learn More
Hampton Research CRYSTAL SCREEN LITE NO16 185ML
SDP
Catalog No. 501621
Encompass
HR2-916-16/Crystal Screen Lite #16/MembFac HT (F04), 185 ml - 0.1 M HEPES sodium pH 7.5, 0.75 M Lithium sulfate monohydrate. Please note items from this supplier are non-returnable.
Catalog No. 50-162-1 Supplier Hampton Research Supplier No. HR291616
HR2-916-16/Crystal Screen Lite #16/MembFac HT (F04), 185 ml - 0.1 M HEPES sodium pH 7.5, 0.75 M Lithium sulfate monohydrate. Please note items from this supplier are non-returnable.