Learn More
Hampton Research CRYSTAL SCREEN LITE NO17 185ML
SDP
Catalog No. 501622
Encompass
HR2-916-17/Crystal Screen Lite #17/MembFac HT (F05), 185 ml - 0.2 M Lithium sulfate monohydrate, 0.1 M TRIS hydrochloride pH 8.5, 15% w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.
Catalog No. 50-162-2 Supplier Hampton Research Supplier No. HR291617
HR2-916-17/Crystal Screen Lite #17/MembFac HT (F05), 185 ml - 0.2 M Lithium sulfate monohydrate, 0.1 M TRIS hydrochloride pH 8.5, 15% w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.