Learn More
Hampton Research CRYSTAL SCREEN LITE NO18 185ML
SDP
Catalog No. 501623
Encompass
HR2-916-18/Crystal Screen Lite #18/MembFac HT (F06), 185 ml - 0.2 M Magnesium acetate tetrahydrate, 0.1 M Sodium cacodylate trihydrate pH 6.5, 10% w/v Polyethylene glycol 8,000. Please note items from this supplier are non-returnable.
Catalog No. 50-162-3 Supplier Hampton Research Supplier No. HR291618
HR2-916-18/Crystal Screen Lite #18/MembFac HT (F06), 185 ml - 0.2 M Magnesium acetate tetrahydrate, 0.1 M Sodium cacodylate trihydrate pH 6.5, 10% w/v Polyethylene glycol 8,000. Please note items from this supplier are non-returnable.