Learn More
Hampton Research CRYSTAL SCREEN LITE NO19 185ML
SDP
Catalog No. 501624
Encompass
HR2-916-19/Crystal Screen Lite #19/MembFac HT (F07), 185 ml - 0.2 M Ammonium acetate, 0.1 M TRIS hydrochloride pH 8.5, 15% v/v 2-Propanol. Please note items from this supplier are non-returnable.
Catalog No. 50-162-4 Supplier Hampton Research Supplier No. HR291619
HR2-916-19/Crystal Screen Lite #19/MembFac HT (F07), 185 ml - 0.2 M Ammonium acetate, 0.1 M TRIS hydrochloride pH 8.5, 15% v/v 2-Propanol. Please note items from this supplier are non-returnable.