Learn More
Hampton Research CRYSTAL SCREEN LITE NO20 185ML
SDP
Catalog No. 501625
Encompass
HR2-916-20/Crystal Screen Lite #20/MembFac HT (F08), 185 ml - 0.2 M Ammonium sulfate, 0.1 M Sodium acetate trihydrate pH 4.6, 12.5% w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.
Catalog No. 50-162-5 Supplier Hampton Research Supplier No. HR291620
HR2-916-20/Crystal Screen Lite #20/MembFac HT (F08), 185 ml - 0.2 M Ammonium sulfate, 0.1 M Sodium acetate trihydrate pH 4.6, 12.5% w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.