Learn More
Hampton Research CRYSTAL SCREEN LITE NO21 185ML
SDP
Catalog No. 501626
Encompass
HR2-916-21/Crystal Screen Lite #21/MembFac HT (F09), 185 ml - 0.2 M Magnesium acetate tetrahydrate, 0.1 M Sodium cacodylate trihydrate pH 6.5, 15% v/v (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.
Catalog No. 50-162-6 Supplier Hampton Research Supplier No. HR291621
HR2-916-21/Crystal Screen Lite #21/MembFac HT (F09), 185 ml - 0.2 M Magnesium acetate tetrahydrate, 0.1 M Sodium cacodylate trihydrate pH 6.5, 15% v/v (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.