Learn More
Hampton Research CRYSTAL SCREEN LITE NO22 185ML
SDP
Catalog No. 501627
Encompass
HR2-916-22/Crystal Screen Lite #22/MembFac HT (F10), 185 ml - 0.2 M Sodium acetate trihydrate, 0.1 M TRIS hydrochloride pH 8.5, 15% w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.
Catalog No. 50-162-7 Supplier Hampton Research Supplier No. HR291622
HR2-916-22/Crystal Screen Lite #22/MembFac HT (F10), 185 ml - 0.2 M Sodium acetate trihydrate, 0.1 M TRIS hydrochloride pH 8.5, 15% w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.