Learn More
Hampton Research CRYSTAL SCREEN LITE NO23 185ML
SDP
Catalog No. 501628
Encompass
HR2-916-23/Crystal Screen Lite #23/MembFac HT (F11), 185 ml - 0.2 M Magnesium chloride hexahydrate, 0.1 M HEPES sodium pH 7.5, 15% v/v Polyethylene glycol 400. Please note items from this supplier are non-returnable.
Catalog No. 50-162-8 Supplier Hampton Research Supplier No. HR291623
HR2-916-23/Crystal Screen Lite #23/MembFac HT (F11), 185 ml - 0.2 M Magnesium chloride hexahydrate, 0.1 M HEPES sodium pH 7.5, 15% v/v Polyethylene glycol 400. Please note items from this supplier are non-returnable.