Learn More
Hampton Research CRYSTAL SCREEN LITE NO24 185ML
SDP
Catalog No. 501629
Encompass
HR2-916-24/Crystal Screen Lite #24/MembFac HT (F12), 185 ml - 0.2 M Calcium chloride dihydrate, 0.1 M Sodium acetate trihydrate pH 4.6, 10% v/v 2-Propanol. Please note items from this supplier are non-returnable.
Catalog No. 50-162-9 Supplier Hampton Research Supplier No. HR291624
HR2-916-24/Crystal Screen Lite #24/MembFac HT (F12), 185 ml - 0.2 M Calcium chloride dihydrate, 0.1 M Sodium acetate trihydrate pH 4.6, 10% v/v 2-Propanol. Please note items from this supplier are non-returnable.