Learn More
Hampton Research CRYSTAL SCREEN LITE NO26 185ML
SDP
Catalog No. 501631
Encompass
HR2-916-26/Crystal Screen Lite #26/MembFac HT (G02), 185 ml - 0.2 M Ammonium acetate, 0.1 M Sodium citrate tribasic dihydrate pH 5.6, 15% v/v (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.
Catalog No. 50-163-1 Supplier Hampton Research Supplier No. HR291626
HR2-916-26/Crystal Screen Lite #26/MembFac HT (G02), 185 ml - 0.2 M Ammonium acetate, 0.1 M Sodium citrate tribasic dihydrate pH 5.6, 15% v/v (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.