Learn More
Hampton Research CRYSTAL SCREEN LITE NO28 185ML
SDP
Catalog No. 501633
Encompass
HR2-916-28/Crystal Screen Lite #28/MembFac HT (G04), 185 ml - 0.2 M Sodium acetate trihydrate, 0.1 M Sodium cacodylate trihydrate pH 6.5, 15% w/v Polyethylene glycol 8,000. Please note items from this supplier are non-returnable.
Catalog No. 50-163-3 Supplier Hampton Research Supplier No. HR291628
HR2-916-28/Crystal Screen Lite #28/MembFac HT (G04), 185 ml - 0.2 M Sodium acetate trihydrate, 0.1 M Sodium cacodylate trihydrate pH 6.5, 15% w/v Polyethylene glycol 8,000. Please note items from this supplier are non-returnable.