Learn More
Hampton Research CRYSTAL SCREEN LITE NO29 185ML
SDP
Catalog No. 501634
Encompass
HR2-916-29/Crystal Screen Lite #29/MembFac HT (G05), 185 ml - 0.1 M HEPES sodium pH 7.5, 0.4 M Potassium sodium tartrate tetrahydrate. Please note items from this supplier are non-returnable.
Catalog No. 50-163-4 Supplier Hampton Research Supplier No. HR291629
HR2-916-29/Crystal Screen Lite #29/MembFac HT (G05), 185 ml - 0.1 M HEPES sodium pH 7.5, 0.4 M Potassium sodium tartrate tetrahydrate. Please note items from this supplier are non-returnable.