Learn More
Hampton Research CRYSTAL SCREEN LITE NO30 185ML
SDP
Catalog No. 501635
Encompass
HR2-916-30/Crystal Screen Lite #30/MembFac HT (G06), 185 ml - 0.2 M Ammonium sulfate, 15% w/v Polyethylene glycol 8,000. Please note items from this supplier are non-returnable.
Catalog No. 50-163-5 Supplier Hampton Research Supplier No. HR291630
HR2-916-30/Crystal Screen Lite #30/MembFac HT (G06), 185 ml - 0.2 M Ammonium sulfate, 15% w/v Polyethylene glycol 8,000. Please note items from this supplier are non-returnable.