Learn More
Hampton Research CRYSTAL SCREEN LITE NO31 185ML
SDP
Catalog No. 501636
Encompass
HR2-916-31/Crystal Screen Lite #31/MembFac HT (G07), 185 ml - 0.2 M Ammonium sulfate, 15% w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.
Catalog No. 50-163-6 Supplier Hampton Research Supplier No. HR291631
HR2-916-31/Crystal Screen Lite #31/MembFac HT (G07), 185 ml - 0.2 M Ammonium sulfate, 15% w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.