Learn More
Hampton Research CRYSTAL SCREEN LITE NO32 185ML
SDP
Catalog No. 501637
Encompass
HR2-916-32/Crystal Screen Lite #32/MembFac HT (G08), 185 ml - 1.0 M Ammonium sulfate. Please note items from this supplier are non-returnable.
Catalog No. 50-163-7 Supplier Hampton Research Supplier No. HR291632
HR2-916-32/Crystal Screen Lite #32/MembFac HT (G08), 185 ml - 1.0 M Ammonium sulfate. Please note items from this supplier are non-returnable.