Learn More
Hampton Research CRYSTAL SCREEN LITE NO33 185ML
SDP
Catalog No. 501638
Encompass
HR2-916-33/Crystal Screen Lite #33/MembFac HT (G09), 185 ml - 2.0 M Sodium formate. Please note items from this supplier are non-returnable.
Catalog No. 50-163-8 Supplier Hampton Research Supplier No. HR291633
HR2-916-33/Crystal Screen Lite #33/MembFac HT (G09), 185 ml - 2.0 M Sodium formate. Please note items from this supplier are non-returnable.