Learn More
Hampton Research CRYSTAL SCREEN LITE NO36 185ML
SDP
Catalog No. 501641
Encompass
HR2-916-36/Crystal Screen Lite #36/MembFac HT (G12), 185 ml - 0.1 M TRIS hydrochloride pH 8.5, 4% w/v Polyethylene glycol 8,000. Please note items from this supplier are non-returnable.
Catalog No. 50-164-1 Supplier Hampton Research Supplier No. HR291636
HR2-916-36/Crystal Screen Lite #36/MembFac HT (G12), 185 ml - 0.1 M TRIS hydrochloride pH 8.5, 4% w/v Polyethylene glycol 8,000. Please note items from this supplier are non-returnable.