Learn More
Hampton Research CRYSTAL SCREEN LITE NO37 185ML
SDP
Catalog No. 501642
Encompass
HR2-916-37/Crystal Screen Lite #37/MembFac HT (H01), 185 ml - 0.1 M Sodium acetate trihydrate pH 4.6, 4% w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.
Catalog No. 50-164-2 Supplier Hampton Research Supplier No. HR291637
HR2-916-37/Crystal Screen Lite #37/MembFac HT (H01), 185 ml - 0.1 M Sodium acetate trihydrate pH 4.6, 4% w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.