Learn More
Hampton Research CRYSTAL SCREEN LITE NO38 185ML
SDP
Catalog No. 501643
Encompass
HR2-916-38/Crystal Screen Lite #38/MembFac HT (H02), 185 ml - 0.1 M HEPES sodium pH 7.5, 0.7 M Sodium citrate tribasic dihydrate. Please note items from this supplier are non-returnable.
Catalog No. 50-164-3 Supplier Hampton Research Supplier No. HR291638
HR2-916-38/Crystal Screen Lite #38/MembFac HT (H02), 185 ml - 0.1 M HEPES sodium pH 7.5, 0.7 M Sodium citrate tribasic dihydrate. Please note items from this supplier are non-returnable.