Learn More
Hampton Research CRYSTAL SCREEN LITE NO39 185ML
SDP
Catalog No. 501644
Encompass
HR2-916-39/Crystal Screen Lite #39/MembFac HT (H03), 185 ml - 0.1 M HEPES sodium pH 7.5, 1.0 M Ammonium sulfate, 2% v/v Polyethylene glycol 400. Please note items from this supplier are non-returnable.
Catalog No. 50-164-4 Supplier Hampton Research Supplier No. HR291639
HR2-916-39/Crystal Screen Lite #39/MembFac HT (H03), 185 ml - 0.1 M HEPES sodium pH 7.5, 1.0 M Ammonium sulfate, 2% v/v Polyethylene glycol 400. Please note items from this supplier are non-returnable.