Learn More
Hampton Research CRYSTAL SCREEN LITE NO41 185ML
SDP
Catalog No. 501646
Encompass
HR2-916-41/Crystal Screen Lite #41/MembFac HT (H05), 185 ml - 0.1 M HEPES sodium pH 7.5, 5% v/v 2-Propanol, 10% w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.
Catalog No. 50-164-6 Supplier Hampton Research Supplier No. HR291641
HR2-916-41/Crystal Screen Lite #41/MembFac HT (H05), 185 ml - 0.1 M HEPES sodium pH 7.5, 5% v/v 2-Propanol, 10% w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.