Learn More
Hampton Research CRYSTAL SCREEN LITE NO43 185ML
SDP
Catalog No. 501648
Encompass
HR2-916-43/Crystal Screen Lite #43/MembFac HT (H07), 185 ml - 15% w/v Polyethylene glycol 1,500. Please note items from this supplier are non-returnable.
Catalog No. 50-164-8 Supplier Hampton Research Supplier No. HR291643
HR2-916-43/Crystal Screen Lite #43/MembFac HT (H07), 185 ml - 15% w/v Polyethylene glycol 1,500. Please note items from this supplier are non-returnable.