Learn More
Hampton Research CRYSTAL SCREEN LITE NO45 185ML
SDP
Catalog No. 501650
Encompass
HR2-916-45/Crystal Screen Lite #45/MembFac HT (H09), 185 ml - 0.2 M Zinc acetate dihydrate, 0.1 M Sodium cacodylate trihydrate pH 6.5, 9% w/v Polyethylene glycol 8,000. Please note items from this supplier are non-returnable.
Catalog No. 50-165-0 Supplier Hampton Research Supplier No. HR291645
HR2-916-45/Crystal Screen Lite #45/MembFac HT (H09), 185 ml - 0.2 M Zinc acetate dihydrate, 0.1 M Sodium cacodylate trihydrate pH 6.5, 9% w/v Polyethylene glycol 8,000. Please note items from this supplier are non-returnable.