Learn More
Hampton Research CRYSTAL SCREEN LITE NO46 185ML
SDP
Catalog No. 501651
Encompass
HR2-916-46/Crystal Screen Lite #46/MembFac HT (H10), 185 ml - 0.2 M Calcium acetate hydrate, 0.1 M Sodium cacodylate trihydrate pH 6.5, 9% w/v Polyethylene glycol 8,000. Please note items from this supplier are non-returnable.
Catalog No. 50-165-1 Supplier Hampton Research Supplier No. HR291646
HR2-916-46/Crystal Screen Lite #46/MembFac HT (H10), 185 ml - 0.2 M Calcium acetate hydrate, 0.1 M Sodium cacodylate trihydrate pH 6.5, 9% w/v Polyethylene glycol 8,000. Please note items from this supplier are non-returnable.