Learn More
Hampton Research CRYSTAL SCREEN LITE NO47 185ML
SDP
Catalog No. 501652
Encompass
HR2-916-47/Crystal Screen Lite #47/MembFac HT (H11), 185 ml - 0.1 M Sodium acetate trihydrate pH 4.6, 1.0 M Ammonium sulfate. Please note items from this supplier are non-returnable.
Catalog No. 50-165-2 Supplier Hampton Research Supplier No. HR291647
HR2-916-47/Crystal Screen Lite #47/MembFac HT (H11), 185 ml - 0.1 M Sodium acetate trihydrate pH 4.6, 1.0 M Ammonium sulfate. Please note items from this supplier are non-returnable.