Learn More
Hampton Research CRYSTAL SCREEN LITE NO48 185ML
SDP
Catalog No. 501653
Encompass
HR2-916-48/Crystal Screen Lite #48/MembFac HT (H12), 185 ml - 0.1 M TRIS hydrochloride pH 8.5, 1.0 M Ammonium phosphate monobasic. Please note items from this supplier are non-returnable.
Catalog No. 50-165-3 Supplier Hampton Research Supplier No. HR291648
HR2-916-48/Crystal Screen Lite #48/MembFac HT (H12), 185 ml - 0.1 M TRIS hydrochloride pH 8.5, 1.0 M Ammonium phosphate monobasic. Please note items from this supplier are non-returnable.