Learn More
Hampton Research CRYSTAL SCREEN LITE NO49 185ML
SDP
Catalog No. 501654
Encompass
HR2-916-49/Crystal Screen Lite #49, 185 ml - 0.5 M Lithium sulfate monohydrate, 2% w/v Polyethylene glycol 8,000. Please note items from this supplier are non-returnable.
Catalog No. 50-165-4 Supplier Hampton Research Supplier No. HR291649
HR2-916-49/Crystal Screen Lite #49, 185 ml - 0.5 M Lithium sulfate monohydrate, 2% w/v Polyethylene glycol 8,000. Please note items from this supplier are non-returnable.