Learn More
Hampton Research CRYSTAL SCREEN LITE NO50 185ML
SDP
Catalog No. 501655
Encompass
HR2-916-50/Crystal Screen Lite #50, 185 ml - 0.5 M Lithium sulfate monohydrate, 7.5% w/v Polyethylene glycol 8,000. Please note items from this supplier are non-returnable.
Catalog No. 50-165-5 Supplier Hampton Research Supplier No. HR291650
HR2-916-50/Crystal Screen Lite #50, 185 ml - 0.5 M Lithium sulfate monohydrate, 7.5% w/v Polyethylene glycol 8,000. Please note items from this supplier are non-returnable.