Learn More
Hampton Research CRYSTAL SCREEN NO06 185ML
SDP
Catalog No. 501415
Encompass
HR2-910-06/Crystal Screen #06 (A6), 185 ml - 0.2 M Magnesium chloride hexahydrate, 0.1 M TRIS hydrochloride pH 8.5, 30% w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.
Catalog No. 50-141-5 Supplier Hampton Research Supplier No. HR291006
HR2-910-06/Crystal Screen #06 (A6), 185 ml - 0.2 M Magnesium chloride hexahydrate, 0.1 M TRIS hydrochloride pH 8.5, 30% w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.