Learn More
Hampton Research CRYSTAL SCREEN NO07 185ML
SDP
Catalog No. 501416
Encompass
HR2-910-07/Crystal Screen #07 (A7), 185 ml - 0.1 M Sodium cacodylate trihydrate pH 6.5, 1.4 M Sodium acetate trihydrate. Please note items from this supplier are non-returnable.
Catalog No. 50-141-6 Supplier Hampton Research Supplier No. HR291007
Add to Cart
Add to Cart
HR2-910-07/Crystal Screen #07 (A7), 185 ml - 0.1 M Sodium cacodylate trihydrate pH 6.5, 1.4 M Sodium acetate trihydrate. Please note items from this supplier are non-returnable.