Learn More
Hampton Research CRYSTAL SCREEN NO08 185ML
SDP
Catalog No. 501417
Encompass
HR2-910-08/Crystal Screen #08 (A8), 185 ml - 0.2 M Sodium citrate tribasic dihydrate, 0.1 M Sodium cacodylate trihydrate pH 6.5, 30% v/v 2-Propanol. Please note items from this supplier are non-returnable.
Catalog No. 50-141-7 Supplier Hampton Research Supplier No. HR291008
HR2-910-08/Crystal Screen #08 (A8), 185 ml - 0.2 M Sodium citrate tribasic dihydrate, 0.1 M Sodium cacodylate trihydrate pH 6.5, 30% v/v 2-Propanol. Please note items from this supplier are non-returnable.