Learn More
Hampton Research CRYSTAL SCREEN NO09 185ML
SDP
Catalog No. 501418
Encompass
HR2-910-09/Crystal Screen #09 (A9), 185 ml - 0.2 M Ammonium acetate, 0.1 M Sodium citrate tribasic dihydrate pH 5.6, 30% w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.
Catalog No. 50-141-8 Supplier Hampton Research Supplier No. HR291009
HR2-910-09/Crystal Screen #09 (A9), 185 ml - 0.2 M Ammonium acetate, 0.1 M Sodium citrate tribasic dihydrate pH 5.6, 30% w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.