Learn More
Hampton Research CRYSTAL SCREEN NO10 185ML
SDP
Catalog No. 501419
Encompass
HR2-910-10/Crystal Screen #10 (A10), 185 ml - 0.2 M Ammonium acetate, 0.1 M Sodium acetate trihydrate pH 4.6, 30% w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.
Catalog No. 50-141-9 Supplier Hampton Research Supplier No. HR291010
HR2-910-10/Crystal Screen #10 (A10), 185 ml - 0.2 M Ammonium acetate, 0.1 M Sodium acetate trihydrate pH 4.6, 30% w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.