Learn More
Hampton Research CRYSTAL SCREEN NO11 185ML
SDP
Catalog No. 501420
Encompass
HR2-910-11/Crystal Screen #11 (A11), 185 ml - 0.1 M Sodium citrate tribasic dihydrate pH 5.6, 1.0 M Ammonium phosphate monobasic. Please note items from this supplier are non-returnable.
Catalog No. 50-142-0 Supplier Hampton Research Supplier No. HR291011
HR2-910-11/Crystal Screen #11 (A11), 185 ml - 0.1 M Sodium citrate tribasic dihydrate pH 5.6, 1.0 M Ammonium phosphate monobasic. Please note items from this supplier are non-returnable.