Learn More
Hampton Research CRYSTAL SCREEN NO13 185ML
SDP
Catalog No. 501422
Encompass
HR2-910-13/Crystal Screen #13 (B1), 185 ml - 0.2 M Sodium citrate tribasic dihydrate, 0.1 M TRIS hydrochloride pH 8.5, 30% v/v Polyethylene glycol 400. Please note items from this supplier are non-returnable.
Catalog No. 50-142-2 Supplier Hampton Research Supplier No. HR291013
HR2-910-13/Crystal Screen #13 (B1), 185 ml - 0.2 M Sodium citrate tribasic dihydrate, 0.1 M TRIS hydrochloride pH 8.5, 30% v/v Polyethylene glycol 400. Please note items from this supplier are non-returnable.