Learn More
Hampton Research CRYSTAL SCREEN NO14 185ML
SDP
Catalog No. 501423
Encompass
HR2-910-14/Crystal Screen #14 (B2), 185 ml - 0.2 M Calcium chloride dihydrate, 0.1 M HEPES sodium pH 7.5, 28% v/v Polyethylene glycol 400. Please note items from this supplier are non-returnable.
Catalog No. 50-142-3 Supplier Hampton Research Supplier No. HR291014
HR2-910-14/Crystal Screen #14 (B2), 185 ml - 0.2 M Calcium chloride dihydrate, 0.1 M HEPES sodium pH 7.5, 28% v/v Polyethylene glycol 400. Please note items from this supplier are non-returnable.