Learn More
Hampton Research CRYSTAL SCREEN NO15 185ML
SDP
Catalog No. 501424
Encompass
HR2-910-15/Crystal Screen #15 (B3), 185 ml - 0.2 M Ammonium sulfate, 0.1 M Sodium cacodylate trihydrate pH 6.5, 30% w/v Polyethylene glycol 8,000. Please note items from this supplier are non-returnable.
Catalog No. 50-142-4 Supplier Hampton Research Supplier No. HR291015
HR2-910-15/Crystal Screen #15 (B3), 185 ml - 0.2 M Ammonium sulfate, 0.1 M Sodium cacodylate trihydrate pH 6.5, 30% w/v Polyethylene glycol 8,000. Please note items from this supplier are non-returnable.