Learn More
Hampton Research CRYSTAL SCREEN NO16 185ML
SDP
Catalog No. 501425
Encompass
HR2-910-16/Crystal Screen #16 (B4), 185 ml - 0.1 M HEPES sodium pH 7.5, 1.5 M Lithium sulfate monohydrate. Please note items from this supplier are non-returnable.
Catalog No. 50-142-5 Supplier Hampton Research Supplier No. HR291016
HR2-910-16/Crystal Screen #16 (B4), 185 ml - 0.1 M HEPES sodium pH 7.5, 1.5 M Lithium sulfate monohydrate. Please note items from this supplier are non-returnable.