Learn More
Hampton Research CRYSTAL SCREEN NO17 185ML
SDP
Catalog No. 501426
Encompass
HR2-910-17/Crystal Screen #17 (B5), 185 ml - 0.2 M Lithium sulfate monohydrate, 0.1 M TRIS hydrochloride pH 8.5, 30% w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.
Catalog No. 50-142-6 Supplier Hampton Research Supplier No. HR291017
HR2-910-17/Crystal Screen #17 (B5), 185 ml - 0.2 M Lithium sulfate monohydrate, 0.1 M TRIS hydrochloride pH 8.5, 30% w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.